missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-Amino-3-benzyloxy-1-propanol hydrochloride, 98+%, Thermo Scientific™
POA - £64.80
Chemical Identifiers
CAS | 58577-95-0 |
---|---|
Molecular Formula | C10H16ClNO2 |
Molecular Weight (g/mol) | 217.693 |
MDL Number | MFCD04112476 |
InChI Key | BJWHQBTUHVIRJJ-HNCPQSOCSA-N |
Synonym | r-2-amino-3-benzyloxy propan-1-ol hydrochloride, l-ser bzl-ol*hcl, 2r-2-amino-3-benzyloxy propan-1-ol hydrochloride, serinol bzl hcl, o-benzyl-l-serinol hydrochloride, r-2-amino-3-benzyloxy-1-propanol hydrochloride salt, 2r-2-amino-3-benzyloxy propan-1-ol-hydrogen chloride 1/1 |
PubChem CID | 17998970 |
IUPAC Name | (2R)-2-amino-3-phenylmethoxypropan-1-ol;hydrochloride |
SMILES | C1=CC=C(C=C1)COCC(CO)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419725
|
Thermo Scientific Alfa Aesar
H52302.03 |
1 g |
£64.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15429725
|
Thermo Scientific Alfa Aesar
H52302.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
58577-95-0 | |
217.693 | |
BJWHQBTUHVIRJJ-HNCPQSOCSA-N | |
17998970 | |
C1=CC=C(C=C1)COCC(CO)N.Cl |
C10H16ClNO2 | |
MFCD04112476 | |
r-2-amino-3-benzyloxy propan-1-ol hydrochloride, l-ser bzl-ol*hcl, 2r-2-amino-3-benzyloxy propan-1-ol hydrochloride, serinol bzl hcl, o-benzyl-l-serinol hydrochloride, r-2-amino-3-benzyloxy-1-propanol hydrochloride salt, 2r-2-amino-3-benzyloxy propan-1-ol-hydrogen chloride 1/1 | |
(2R)-2-amino-3-phenylmethoxypropan-1-ol;hydrochloride |
Specifications
58577-95-0 | |
MFCD04112476 | |
BJWHQBTUHVIRJJ-HNCPQSOCSA-N | |
(2R)-2-amino-3-phenylmethoxypropan-1-ol;hydrochloride | |
17998970 | |
≥98% |
C10H16ClNO2 | |
r-2-amino-3-benzyloxy propan-1-ol hydrochloride, l-ser bzl-ol*hcl, 2r-2-amino-3-benzyloxy propan-1-ol hydrochloride, serinol bzl hcl, o-benzyl-l-serinol hydrochloride, r-2-amino-3-benzyloxy-1-propanol hydrochloride salt, 2r-2-amino-3-benzyloxy propan-1-ol-hydrogen chloride 1/1 | |
C1=CC=C(C=C1)COCC(CO)N.Cl | |
217.693 | |
217.7 | |
(R)-2-Amino-3-benzyloxy-1-propanol hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures