missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-2-Amino-3-benzyloxy-1-propanol, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS | 58577-87-0 |
| Melting Point | 34°C to 37°C |
| Boiling Point | 307°C |
| Flash Point | 113°C |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass Bottle |
| Molecular Formula | C10H15NO2 |
| Linear Formula | C6H5CH2OCH2CH(NH2)CH2OH |
| Quantity | 1 g |
| Synonym | r-2-amino-3-benzyloxy propan-1-ol, h-serinol bzl, r-+-2-amino-3-benzyloxy-1-propanol, 2r-2-amino-3-benzyloxy propan-1-ol, 2-amino-3-benzyloxy-1-propanol, 1-propanol, 2-amino-3-phenylmethoxy-, 2r, r-2-amino-3-benzyloxy-1-propanol, serinol benzyl ether, ambotzhal1055, r-2-amino-3-benzyloxypropan-1-ol |
| Show More |
Safety and Handling
Product Identifier
- (R)-(+)-2-Amino-3-benzyloxy-1-propanol
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?