missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2-Amino-1-propanol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (2R)-2-Aminopropan-1-ol |
| CAS | 35320-23-1 |
| Molecular Formula | C3 H9 N O |
| Quantity | 1 g |
| Synonym | 1-Propanol, 2-amino-, (2R)-, 1-Propanol, 2-amino-, (R)-, (2R)-2-Amino-1-propanol, (-)-2-Amino-1-propanol, (-)-2-Aminopropanol, (-)-Alaninol, (2R)-2-Amino-1-hydroxypropane, (2R)-2-Amino-1-propanol, (2R)-2-Aminopropan-1-ol, (R)-(-)-2-Amino-1-propanol, (R)-(-)-2-Aminopropanol, (R)-2-Amino-1-propanol, (R)-2-Aminopropan-1-ol, (R)-2-Aminopropanol, (R)-Alaninol, 2R-Aminopropanol, D-2-Amino-1-propanol, D-Alaninol, R-(-)-Alaninol, [(R)-2-Hydroxy-1-methylethyl]amine |
| InChI Formula | InChI=1S/C3H9NO/c1-3(4)2-5/h3,5H,2,4H2,1H3/t3-/m1/s1 |
| SMILES | C[C@@H](N)CO |
| IUPAC Name | (2R)-2-aminopropan-1-ol |
| Molecular Weight (g/mol) | 75.1097 |
| Formula Weight | 75.0684 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction