missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-butanol, 98%

Product Code. 11430708
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430708 5 g 5g
11440708 25 g 25g
11450708 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430708 Supplier Thermo Scientific Alfa Aesar Supplier No. B25212.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5856-63-3 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5856-63-3
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00064419
InChI Key JCBPETKZIGVZRE-UHFFFAOYNA-N
Synonym r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r
PubChem CID 2723856
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(N)CO

Specifications

Density 0.947
pH 11.1
Boiling Point 176°C to 178°C
Flash Point 89°C (192°F)
Odor Amine-like
Refractive Index 1.4525
Quantity 5 g
UN Number UN2735
Beilstein 1718929
Sensitivity Air sensitive; Hygroscopic
Optical Rotation −10° (Neat)
Formula Weight 89.14
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Amino-1-butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.