missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]-3-methylbutanamide, TRC

Product Code. 30401018
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401018 250 mg
30390430 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401018 Supplier Toronto Research Chemicals Supplier No. TRCF591505250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210529-62-7
Molecular Formula C11H13FN4O5
Molecular Weight (g/mol) 300.24
Synonym (2R)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]-3-methylbutanamide
SMILES CC(C)[C@@H](Nc1cc(F)c(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H13FN4O5/c1-5(2)10(11(13)17)14-7-3-6(12)8(15(18)19)4-9(7)16(20)21/h3-5,10,14H,1-2H3,(H2,13,17)/t10-/m1/s1
Formula Weight 300.087
Chemical Name or Material (R)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]-3-methylbutanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.