missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid, TRC

Product Code. 30393771
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393771 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393771 Supplier Toronto Research Chemicals Supplier No. TRCH9497155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid
CAS 94050-90-5
Molecular Formula C9H10O4
Quantity 5 g
Synonym (2R)-2-(4-Hydroxyphenoxy)propanoic Acid, (+)-2-(4-Hydroxyphenoxy)propionic Acid, (R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid, (R)-2-(4-Hydroxyphenoxy)propanoic Acid, (R)-2-(4-Hydroxyphenoxy)propanoic Acid, (R)-2-(4-Hydroxyphenoxy)propionic Acid, (R)-2-(p-Hydroxyphenoxy)propionic Acid, D-2-(4-Hydroxyphenoxy)propionic Acid, HPPA
InChI Formula InChI=1S/C9H10O4/c1-6(9(11)12)13-8-4-2-7(10)3-5-8/h2-6,10H,1H3,(H,11,12)/t6-/m1/s1
SMILES C[C@@H](Oc1ccc(O)cc1)C(=O)O
IUPAC Name (2R)-2-(4-hydroxyphenoxy)propanoic acid
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.