missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2,3-O-Isopropylideneglycerol, 96%, Thermo Scientific Chemicals

Product Code. 11446226
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446226 5 g 5g
11436226 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446226 Supplier Thermo Scientific Alfa Aesar Supplier No. B23037.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14347-78-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00003213
InChI Key RNVYQYLELCKWAN-RXMQYKEDSA-N
Synonym r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol
PubChem CID 736056
IUPAC Name [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(OCC(O1)CO)C

Specifications

Density 1.066
Boiling Point 82°C to 83°C (14 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.435
Quantity 5 g
Beilstein 80117
Merck Index 14,5213
Optical Rotation −11.5° (c=5 in Methanol)
Formula Weight 132.18
Percent Purity 96%
Chemical Name or Material (R)-(-)-2,3-O-Isopropylideneglycerol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.