missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Stearate, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Stearate |
| Analyte or Component Names | (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Stearate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H46 O4 |
| InChI Formula | InChI=1S/C24H46O4/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(25)26-20-22-21-27-24(2,3)28-22/h22H,4-21H2,1-3H3/t22-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OC[C@H]1COC(C)(C)O1 |
| IUPAC Name | [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl octadecanoate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 398.62 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?