missing translation for 'onlineSavingsMsg'
Learn More

(R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Stearate, TRC

Product Code. 30376019
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376019 100 mg 100mg
30379781 500 mg 500mg
30384687 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376019 Supplier Toronto Research Chemicals Supplier No. TRCD579030100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Stearate
Analyte or Component Names (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Stearate
Shipping Condition Room Temperature
Molecular Formula C24 H46 O4
InChI Formula InChI=1S/C24H46O4/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(25)26-20-22-21-27-24(2,3)28-22/h22H,4-21H2,1-3H3/t22-/m0/s1
Solution Type Neat
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@H]1COC(C)(C)O1
IUPAC Name [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl octadecanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 398.62
Quantity 100 mg
Formula Weight 398.34
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.