missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, CTH-(R)-BINAM, 95%, Thermo Scientific™

Product Code. 11465420
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
This item is not returnable. View return policy

Product Code. 11465420

Brand: Thermo Scientific Alfa Aesar 044620.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Asymmetric hydrogenation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208248-67-3
Molecular Formula C44H42N2P2
Molecular Weight (g/mol) 660.78
MDL Number MFCD04974231
InChI Key GTPWPBWUIWYZCB-UHFFFAOYSA-N
Synonym s---2,2'-bis n-diphenylphosphinoamino-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, cth-s-binam, r-+-2,2'-bis n-diphenylphosphinoamino-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, phosphinous amide,n,n'-1s-5,5',6,6',7,7',8,8'-octahydro 1,1'-binaphthalene-2,2'-diyl bis p,p-diphenyl-9ci, 5,5'-bi 6-diphenylphosphinoamino tetralin, s---2,2'-bis n-diphenylphosphinoamino-5,5'6,6'7,7',8,8'-octahydro-1,1'-binaphthyl, r-+-2,2'-bis n-diphenylphosphinoamino-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, cth-r-binam, s---2,2'-bis n-diphenylphosphinoamino-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, s---2,2'-bis n-diphenylphosphinoamino-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, cth-s-bi, n-2'-diphenylphosphanyl amino-5h,5'h,6h,6'h,7h,7'h,8h,8'h-1,1'-binaphthalen-2-yl-1,1-diphenylphosphanamine
PubChem CID 10931066
IUPAC Name N-diphenylphosphanyl-1-[2-(diphenylphosphanylamino)-5,6,7,8-tetrahydronaphthalen-1-yl]-5,6,7,8-tetrahydronaphthalen-2-amine
SMILES C1CCC2=C(C(NP(C3=CC=CC=C3)C3=CC=CC=C3)=CC=C2C1)C1=C(NP(C2=CC=CC=C2)C2=CC=CC=C2)C=CC2=C1CCCC2

Specifications

Quantity 0.5 g
Formula Weight 660.78
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (R)- (+)-2,2'-Bis (N-diphenylphosphinoamino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, CTH-(R)-BINAM
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.