missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 97%, Thermo Scientific Chemicals

Product Code. 15467895
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467895 1 g 1g
15477895 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467895 Supplier Thermo Scientific Alfa Aesar Supplier No. H31791.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173831-50-0
Molecular Formula C24H22O4
Molecular Weight (g/mol) 374.436
MDL Number MFCD03788935
InChI Key YIAQRNNJNMLGTP-UHFFFAOYSA-N
Synonym r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
PubChem CID 10959708
IUPAC Name 2-(methoxymethoxy)-1-[2-(methoxymethoxy)naphthalen-1-yl]naphthalene
SMILES COCOC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OCOC

Specifications

Melting Point 101°C to 105°C
Quantity 1 g
Optical Rotation +94.5° (C=1 in THF)
Formula Weight 374.44
Percent Purity 97%
Chemical Name or Material (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.