missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%

Product Code. 15415485
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415485 250 mg 250mg
15405485 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415485 Supplier Thermo Scientific Alfa Aesar Supplier No. H60684.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1365531-93-6 | C52H60O8P2 | 874.992 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-93-6
Molecular Formula C52H60O8P2
Molecular Weight (g/mol) 874.992
MDL Number MFCD19443626
InChI Key PKHREKHNGLJKHV-UHFFFAOYSA-N
Synonym 2'-bis 4-methoxy-3,5-dimethylphenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl bis 4-methoxy-3,5-dimethylphenyl phosphane, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-dimethyl-4-methoxyphenyl phosphino biphenyl, r-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 4-methoxy-3,5-dimethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 4-methoxy-3,5-dimethylphenyl phosphine
PubChem CID 87110691
IUPAC Name [2-[2-bis(4-methoxy-3,5-dimethylphenyl)phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis(4-methoxy-3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1OC)C)P(C2=CC(=C(C(=C2)C)OC)C)C3=CC(=CC(=C3C4=C(C=C(C=C4P(C5=CC(=C(C(=C5)C)OC)C)C6=CC(=C(C(=C6)C)OC)C)OC)OC)OC)OC

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 874.98
Percent Purity ≥97%
Chemical Name or Material (R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.