missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(1-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 10601325 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10601325 1 g Glass bottle 1g
10331853 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10601325 Supplier Thermo Scientific Acros Supplier No. 382860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27911-63-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD04972322
InChI Key PPHIIIRFJKDTLG-ZCFIWIBFSA-N
Synonym r-1-pyridin-2-yl ethanol, r-2-1-hydroxyethyl pyridine, r-1-2-pyridyl ethanol, 1r-1-pyridin-2-yl ethan-1-ol, r-alpha-methyl-2-pyridinemethanol, 1r-1-pyridin-2-ylethanol, 1r-1-pyridin-2-yl ethanol, pubchem5699, r-1-pyridin-2-yl-ethanol, 1r-1-2-pyridyl ethanol
PubChem CID 642847
IUPAC Name (1R)-1-pyridin-2-ylethanol
SMILES CC(C1=CC=CC=N1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5270 to 1.529
Quantity 1 g
Specific Rotation Condition +24° (31°C c=1 toluene)
Specific Rotation + 24
Formula Weight 123.15
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material (R)-2-(1-Hydroxyethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • (R)-2-(1-Hydroxyethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.