missing translation for 'onlineSavingsMsg'
Learn More

(R)-10-Monohydroxy-10,11-dihydro Carbamazepine, TRC

Product Code. 30179907
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179907 25 mg 25mg
30180331 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179907 Supplier Toronto Research Chemicals Supplier No. TRCM54650025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104746-03-4
Molecular Formula C15H14N2O2
Molecular Weight (g/mol) 254.28
Synonym 5H-Dibenz[b,f]azepine-5-carboxamide, 10,11-dihydro-10-hydroxy-, (10R)-, 5H-Dibenz[b,f]azepine-5-carboxamide, 10,11-dihydro-10-hydroxy-, (R)-, (10R)-10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, (R)-(-)-10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, (R)-(-)-10,11-Dihydro-10-hydroxy-5H-dibenzo[b,f]azepine-5-carboxamide, (R)-Licarbazepine, CGP 13698, (R)-10-Hydroxy-10,11-dihydrocarbamazepine
IUPAC Name (5R)-5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
SMILES NC(=O)N1c2ccccc2C[C@@H](O)c3ccccc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)/t14-/m1/s1
Formula Weight 254.1055
Percent Purity >95%
Chemical Name or Material R-10-Monohydroxy-dihydro-carbamazepin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.