missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals

Product Code. 10038981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10038981 500 mg Glass bottle 500mg
10670893 100 mg Glass bottle 100mg
10094703 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10038981 Supplier Thermo Scientific Acros Supplier No. 370675000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155806-35-2
Molecular Formula C38H50FeOP2
Molecular Weight (g/mol) 640.61
MDL Number MFCD00800284
InChI Key YOYYLEHTACDQJL-BHACGSQPNA-N
Synonym 2r-1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino ferrocene, r,s-josiphos, r-s-ppf-p cy 2, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine ethanol adduct, r-1-sp-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, ferrocene, 1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r, ferrocene,1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r
PubChem CID 10984818
IUPAC Name cyclopentane;dicyclohexyl-[(1R)-1-(2-diphenylphosphanylcyclopentyl)ethyl]phosphane;iron
SMILES [Fe].CCO.c1cccc1.C[C@@H](P(C1CCCCC1)C1CCCCC1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 500 mg
Specific Rotation Condition - 350.00 (20.00°C c=0.4,CHCl3)
Formula Weight 640.61
Percent Purity ≥97.0% (Total base) contains approx. 1 M of ethanol (Total base) of crystallization (Total base)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (R)-(-)-1-[(S)-2-Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.