missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylpropyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 15419159
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15419159

Brand: Thermo Scientific Alfa Aesar L20318.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 745784-00-3
Boiling Point 128°C to 129°C (9 mmHg)
Molecular Formula C10H11NS
Refractive Index 1.5717
MDL Number MFCD05664118
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym r-+-1-phenylpropyl isothiocyanate, 1r-1-isothiocyanatopropyl benzene, 1r-1-phenylpropanisothiocyanate, r-+-1-phenylpropylisothiocyanate
InChI Key KLNMIWGOGGBFNV-UHFFFAOYNA-N
SMILES CCC(N=C=S)C1=CC=CC=C1
IUPAC Name [(1R)-1-isothiocyanatopropyl]benzene
Molecular Weight (g/mol) 177.27
PubChem CID 7018279
Formula Weight 177.27
Percent Purity 97%
Chemical Name or Material (R)-(+)-1-Phenylpropyl isothiocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.