missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-(-)-1-Phenylethyl isothiocyanate, 95%

Product Code. 15499149
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499149 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499149 Supplier Alfa Aesar™ Supplier No. L20315.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 24277-44-9
Density 1.06
Boiling Point 132°C to 133°C (20 mmHg)
Flash Point 103°C (217°F)
Odor Irritating
Molecular Formula C9H9NS
Refractive Index 1.581
MDL Number MFCD00135478
Quantity 1 g
UN Number UN2922
Beilstein 6476425
Sensitivity Moisture sensitive
Synonym r-1-isothiocyanatoethyl benzene, 1r-1-isothiocyanatoethyl benzene, r---1-phenylethyl isothiocyanate, d-alpha-methylbenzyl isothiocyanate, d-a-methylbenzylisothiocyanate, d-a-methylbenzyl isothiocyanate, d--methylbenzyl isothiocyanate, r-methyl benzyl isothiocyanate, r-1-phenylethyl isothiocyanate, --1-phenylethyl isothiocyanate
InChI Key QQCJPTVZIZVKEZ-UHFFFAOYNA-N
Optical Rotation −17° (c=10 in Chloroform)
SMILES CC(N=C=S)C1=CC=CC=C1
IUPAC Name [(1R)-1-isothiocyanatoethyl]benzene
Molecular Weight (g/mol) 163.24
PubChem CID 7015180
Formula Weight 163.24
Percent Purity 95%
Chemical Name or Material (R)-(-)-1-Phenylethyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.