missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethyl isocyanate, 99%

Product Code. 11358365
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358365 5 g 5g
11348365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358365 Supplier Thermo Scientific Alfa Aesar Supplier No. L10305.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 33375-06-3 | C9H9NO | 147.177 g/mol

(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.

Solubility
Decomposes in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33375-06-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00063015
InChI Key JJSCUXAFAJEQGB-MRVPVSSYSA-N
Synonym r-+-1-phenylethyl isocyanate, 1r-1-isocyanatoethyl benzene, r-+-alpha-methylbenzyl isocyanate, benzene, 1r-1-isocyanatoethyl, isocyanic acid r-+-1-phenylethyl ester, isocyanic acid r-+-alpha-methylbenzyl ester, isocyanic acid-r-+-methylbenzyl ester, 1-isocyanatoethyl benzene #, +-alpha-methylbenzyl isocyanate, pubchem8096
PubChem CID 7018262
IUPAC Name [(1R)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

Density 1.045
Boiling Point 55°C to 56°C (2.5 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.5124
Quantity 5 g
UN Number UN2206
Beilstein 3196930
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Optical Rotation +10.5° (Neat)
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material (R)-(+)-1-Phenylethyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.