missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-Methyl-3-pyrrolidinol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | (R)-1-Methyl-3-pyrrolidinol |
| CAS | 104641-60-3 |
| Molecular Formula | C5H11NO |
| Quantity | 5 g |
| Synonym | (R)-1-Methyl-3-pyrrolidinol, (3R)-1-Methylpyrrolidin-3-ol, (R)-1-Methyl-3-pyrrolidinol, (R)-N-Methyl-3-pyrrolidinol |
| InChI Formula | InChI=1S/C5H11NO/c1-6-3-2-5(7)4-6/h5,7H,2-4H2,1H3/t5-/m1/s1 |
| SMILES | O[C@H]1CN(C)CC1 |
| IUPAC Name | (3R)-1-methylpyrrolidin-3-ol |
| Molecular Weight (g/mol) | 101.15 |
| Formula Weight | 101.0841 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction