missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Cyclohexylethylamine, ChiPros 98%, ee 94+%

Product Code. 11319595
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319595 5 g 5g
11309595 1 g 1g
11329595 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319595 Supplier Thermo Scientific Alfa Aesar Supplier No. L19051.06
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5913-13-3 | C8H17N | 127.23 g/mol

(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.

Solubility
Miscible with acetone, chloroform,dimethyl sulfoxide, ethanol, methanol, ether, petroleum ether and dichloromethane. Insoluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5913-13-3
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00043338
InChI Key XBWOPGDJMAJJDG-UHFFFAOYNA-N
Synonym r---1-cyclohexylethylamine, r-1-cyclohexylethylamine, 1r-1-cyclohexylethanamine, r-1-cyclohexylethanamine, 1r-1-cyclohexylethylamine, chembl19290, 1r-1-cyclohexylethan-1-amine, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, r---cyclohexylethylamine
PubChem CID 10997046
SMILES CC(N)C1CCCCC1

Specifications

Density 0.866
pH 11
Boiling Point 177°C to 178°C
Flash Point 51°C (123°F)
Odor Amine-like
Refractive Index 1.462
Quantity 5 g
UN Number UN2733
Beilstein 2935070
Sensitivity Air Sensitive
Solubility Information Miscible with acetone,chloroform,dimethyl sulfoxide,ethanol,methanol,ether,petroleum ether and dichloromethane. Insoluble in water.
Optical Rotation −4° (Neat)
IUPAC Name 1-cyclohexylethan-1-amine
Formula Weight 127.23
Percent Purity 98%
Assay ee ≥94%
Chemical Name or Material (R)-(-)-1-Cyclohexylethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.