missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Boc-3-bromopyrrolidine, 95%

Product Code. 15493748
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15493748

Brand: Thermo Scientific Alfa Aesar H57228.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 569660-97-5 | C9H16BrNO2 | 250.136 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569660-97-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.136
MDL Number MFCD17214727
InChI Key QJTKPXFJOXKUEY-SSDOTTSWSA-N
Synonym r-tert-butyl 3-bromopyrrolidine-1-carboxylate, r-3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3r-3-bromopyrrolidine-1-carboxylate, r---1-boc-3-bromopyrrolidine, s-+-1-boc-3-bromopyrrolidine, 1-boc-r-3-bromopyrrolidine, tert-butyl r-3-bromopyrrolidine-1-carboxylate, r-3-bromo-pyrrolidine-1-carboxylic acid tert-but, r-3-bromopyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 34179476
IUPAC Name tert-butyl (3R)-3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)Br

Specifications

Quantity 1 g
UN Number UN3077
Optical Rotation −33° (c=0.8 in Chloroform)
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (R)-(-)-1-Boc-3-bromopyrrolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.