missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Boc-3-aminopyrrolidine, 97%

Product Code. 11387619
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387619 1 g 1g
11397619 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387619 Supplier Thermo Scientific Alfa Aesar Supplier No. H28188.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 147081-49-0 | C9H18N2O2 | 186.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147081-49-0
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD03419272
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-N
Synonym r-+-1-boc-3-aminopyrrolidine, r-1-boc-3-aminopyrrolidine, r-3-amino-1-n-boc-pyrrolidine, r-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3r-3-aminopyrrolidine-1-carboxylate, r-bocap, r-+-n-boc-3-aminopyrrolidine, 3r-3-amino-1-tert-butoxycarbonyl pyrrolidine, r-n-boc-3-aminopyrrolidine, r-3-amino-n-boc-pyrrolidine
PubChem CID 854070
IUPAC Name tert-butyl (3R)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)N

Specifications

Density 1.098
Boiling Point 243°C to 244°C
Flash Point 91°C (196°F)
Refractive Index 1.472
Quantity 1 g
UN Number UN2735
Sensitivity Air sensitive
Optical Rotation +3.5° (c=1 in Chloroform)
Formula Weight 186.25
Percent Purity 97%
Chemical Name or Material (R)-(+)-1-Boc-3-aminopyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.