missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Benzyl-3-aminopyrrolidine, 99%, ee 99%

Product Code. p-7047734
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11365515 1 g 1g
11375515 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11365515

Brand: Thermo Scientific Alfa Aesar L19690.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114715-39-8 | C11H16N2 | 176.26 g/mol

(R)-(-)-1-Benzyl-3-aminopyrrolidine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Benzyl-3-aminopyrrolidine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114715-39-8
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00082638
InChI Key HBVNLKQGRZPGRP-LLVKDONJSA-N
Synonym r---1-benzyl-3-aminopyrrolidine, r-1-benzyl-3-aminopyrrolidine, r-1-benzylpyrrolidin-3-amine, 3r---1-benzyl-3-aminopyrrolidine, 3r-1-benzylpyrrolidin-3-amine, r-3-amino-1-n-benzyl-pyrrolidine, r-n-benzyl-3-aminopyrrolidine, 3-pyrrolidinamine, 1-phenylmethyl-, 3r, r-3-amino-1-benzylpyrrolidine, 3r-1-benzyl-3-pyrrolidinamine
PubChem CID 1519354
IUPAC Name (3R)-1-benzylpyrrolidin-3-amine
SMILES N[C@@H]1CCN(CC2=CC=CC=C2)C1

Specifications

Density 1.02
Boiling Point 101°C to 103°C (3 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.543
Quantity 1 g
UN Number UN2735
Beilstein 6477592
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Optical Rotation −2° (Neat)
Formula Weight 176.26
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-(-)-1-Benzyl-3-aminopyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.