missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 98%, Thermo Scientific™

Product Code. 11339665
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339665 1 g 1g
11349665 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339665 Supplier Thermo Scientific Alfa Aesar Supplier No. L19120.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(+)-1-(4-Fluorophenyl)ethylamine, is used as a pharmaceutical intermediate and as a chemical in organic synthesis

Solubility
Sparingly Soluble in water (1.5E-3 g/L) (25°C).

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374898-01-8
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03093090
InChI Key QGCLEUGNYRXBMZ-ZCFIWIBFSA-N
Synonym r-1-4-fluorophenyl ethanamine, r-1-4-fluorophenyl ethylamine, 1r-1-4-fluorophenyl ethanamine, r-1-4-fluorophenyl-ethylamine, r-+-1-4-fluorophenyl ethylamine, 1r-+-1-4-fluorophenyl ethylamine, pubchem15219, 1r-1-4-fluorophenyl ethylamine, r-1-4-fluoro-phenyl ethylamine, 4-fluoro-alpha-r-methylbenzylamine
PubChem CID 6950187
IUPAC Name (1R)-1-(4-fluorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)F)N

Specifications

Melting Point -30°C
Density 1.03
Boiling Point 76°C (22 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.502
Quantity 1 g
UN Number UN2735
Beilstein 8614058
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble in water (1.5E-3g/L) (25°C).
Optical Rotation +33° (Neat)
Formula Weight 139.17
Percent Purity 99%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-(4-Fluorophenyl)ethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.