missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(4-Chlorophenyl)ethylamine, 97%, 98% ee

Product Code. 10761554 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10761554 5 g Glass bottle 5g
10390374 1 g Glass bottle 1g
10208903 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10761554 Supplier Thermo Scientific Acros Supplier No. 370650050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 27298-99-3 | C8H10ClN | 155.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27298-99-3
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD00671639
InChI Key PINPOEWMCLFRRB-ZCFIWIBFSA-N
Synonym r-1-4-chlorophenyl ethylamine, r-1-4-chlorophenyl ethanamine, 1r-1-4-chlorophenyl ethanamine, r-+-1-4-chlorophenyl ethylamine, 1r-1-4-chlorophenyl ethan-1-amine, benzenemethanamine, 4-chloro-alpha-methyl-, alphar, pubchem15222, r-4-chloro-, a-methylbenzylamine, 1r-1-4-chlorophenyl ethylamine, r-1-4-chloro-phenyl ethylamine
PubChem CID 1715226
IUPAC Name (1R)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N

Specifications

Color Colorless to Yellow
Density 1.1150g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5410 to 1.5430
Assay Percent Range 96% min. (GC)
Specific Gravity 1.115
Linear Formula ClC6H4CH(CH3)NH2
Boiling Point 232.0°C
Solubility Information Solubility in water: moderately soluble
Formula Weight 155.63
Percent Purity 97% (98% ee)
Chemical Name or Material (R)-1-(4-Chlorophenyl)ethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-1-(4-Chlorophenyl)ethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.