missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, 97%, 99+% ee, Thermo Scientific™

Product Code. 10249523 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249523 25 g Glass bottle 25g
10242382 1 g Glass bottle 1g
10713223 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249523 Supplier Thermo Scientific Acros Supplier No. 367170250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 64265-77-6
Molecular Formula C8H11BrClN
Molecular Weight (g/mol) 236.54
MDL Number MFCD02259386
InChI Key BQCAANUXMMQVAY-UHFFFAOYNA-N
Synonym r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1
PubChem CID 2734642
IUPAC Name (1R)-1-(4-bromophenyl)ethanamine;hydrochloride
SMILES Cl.CC(N)C1=CC=C(Br)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 238°C to 245°C
Color White to Yellow
Enantiomeric Excess 99.0% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with alcohols
Formula Weight 236.54
Percent Purity 97%,99+% ee
Physical Form Powder
Chemical Name or Material (R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, ≥99%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.