missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethylamine N-Acetyl-D-leucine Salt, TRC

Product Code. 30372404
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372404 250 mg 250mg
30383144 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372404 Supplier Toronto Research Chemicals Supplier No. TRCE791715250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethylamine N-Acetyl-D-leucine Salt
CAS 608142-28-5
Molecular Formula C12H19NO4S • C8H15NO3
Quantity 250 mg
Synonym (R)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethan-1-amine Acetyl-D-leucine Salt
InChI Formula InChI=1S/C12H19NO4S.C8H15NO3/c1-4-17-12-7-9(5-6-11(12)16-2)10(13)8-18(3,14)15;1-5(2)4-7(8(11)12)9-6(3)10/h5-7,10H,4,8,13H2,1-3H3;5,7H,4H2,1-3H3,(H,9,10)(H,11,12)/t10-;7-/m01/s1
SMILES CCOC1=C(OC)C=CC([C@@H](N)CS(=O)(C)=O)=C1.CC(C)C[C@@H](NC(C)=O)C(O)=O
IUPAC Name (R)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethan-1-amine acetyl-D-leucinate
Molecular Weight (g/mol) 446.56
Formula Weight 446.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.