missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1,3-Butanediol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | (R)-(-)-1,3-Butanediol |
| CAS | 6290-03-5 |
| Molecular Formula | C4 H10 O2 |
| Quantity | 10 g |
| Synonym | (3R)-1,3-Butanediol, (-)-1,3-Butanediol, (R)-(-)-1,3-Butanediol, (R)-1,3-Butanediol, L-Butane-1,3-diol |
| InChI Formula | InChI=1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3/t4-/m1/s1 |
| SMILES | C[C@@H](O)CCO |
| IUPAC Name | (3R)-butane-1,3-diol |
| Molecular Weight (g/mol) | 90.121 |
| Formula Weight | 90.068 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction