missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,3-Butanediol, 98%

Product Code. 11456167
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11456167

Brand: Thermo Scientific Alfa Aesar B25124.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6290-03-5 | C8H15NO4 | 189.21 g/mol

(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-03-5
MDL Number MFCD00064277
ChEBI CHEBI:52687

Specifications

Density 1.005
Boiling Point 203°C to 204°C
Flash Point 121°C (249°F)
Odor Weak
Molecular Formula C4H10O2
Refractive Index 1.44
Quantity 5 g
Beilstein 1718944
Sensitivity Hygroscopic
Synonym r---1,3-butanediol, r-butane-1,3-diol, 3r-butane-1,3-diol, r-1,3-butanediol, 1,3-butanediol, 3r, r---1,3-butylene glycol, r---butane-1,3-diol, 1,3-butanediol, r
Solubility Information Fully miscible in water.
InChI Key PUPZLCDOIYMWBV-UHFFFAOYNA-N
Optical Rotation −30° (Neat)
SMILES CC(CCO)O
IUPAC Name (3R)-butane-1,3-diol
Molecular Weight (g/mol) 90.12
PubChem CID 637497
Formula Weight 90.12
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,3-Butanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.