missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(3-Bromophenyl)ethylamine, ChiPros™, 99%, ee 98+%

Product Code. 11486590
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11486590

Brand: Thermo Scientific Alfa Aesar H27636.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 176707-77-0 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176707-77-0
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00671637
InChI Key LIBZHYLTOAGURM-ZCFIWIBFSA-N
Synonym r-1-3-bromophenyl ethanamine, r-1-3-bromophenyl ethylamine, 1r-1-3-bromophenyl ethanamine, 1r-1-3-bromophenyl ethan-1-amine, s-3-bromo-alpha-methylbenzylamine, 1s-1-3-bromophenyl ethylamine, pubchem14235, s---3-bromo alpha-methylbenzylamine, r-3-bromo-, a-methylbenzylamine, r-1-3-bromophenyl ethaneamine
PubChem CID 22831503
IUPAC Name (1R)-1-(3-bromophenyl)ethanamine
SMILES CC(C1=CC(=CC=C1)Br)N

Specifications

Density 1.33
Boiling Point 96°C (4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Optical Rotation +22° (Neat)
Formula Weight 200.08
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (R)-1-(3-Bromophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.