missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol, 98%

Product Code. 15407408
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15407408 Supplier Thermo Scientific Alfa Aesar Supplier No. H60893.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 127852-28-2 | C10H8F6O | 258.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127852-28-2
Melting Point 53°C to 58°C
Color White
Molecular Formula C10H8F6O
MDL Number MFCD03093010
Quantity 5 g
Synonym r-1-3,5-bis trifluoromethyl phenyl ethanol, 1r-1-3,5-bis trifluoromethyl phenyl ethanol, r-1-3,5-bis-trifluoromethyl-phenyl-ethanol, r-1-3,5-bis-trifluoromethylphenyl ethanol, r-1-3,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-3,5-bis trifluoromethyl-, ar, zlchem 1281
InChI Key MMSCIQKQJVBPIR-UHFFFAOYNA-N
SMILES CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
IUPAC Name (1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol) 258.16
PubChem CID 2779031
Formula Weight 258.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.