missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%

Product Code. 11302749
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302749 1 g 1g
11312749 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302749 Supplier Thermo Scientific Alfa Aesar Supplier No. H27669.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 68285-23-4 | C9H13NO | 151.21 g/mol

The derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68285-23-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00671657
InChI Key VENQOHAPVLVQKV-UHFFFAOYNA-N
Synonym r-1-2-methoxyphenyl ethanamine, r-1-2-methoxyphenyl ethylamine, 1r-1-2-methoxyphenyl ethanamine, 1r-1-2-methoxyphenyl ethan-1-amine, r-2-methoxy-, a-methylbenzylamine, 1r-1-2-methoxyphenyl ethylamine, r-2-methoxy-alpha-methylbenzylamine, r-+-2-methoxy a-methylbenzylamine, r-1-2-methoxyphenyl ethylamine, chipros, r-2-methoxy-alpha-methylbenzylamine, chipros r , produced by basf
PubChem CID 853083
IUPAC Name (1R)-1-(2-methoxyphenyl)ethanamine
SMILES COC1=CC=CC=C1C(C)N

Specifications

Boiling Point 118°C (17 mmHg)
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 151.21
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (R)-1-(2-Methoxyphenyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.