missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-1-(2-Chlorophenyl)ethanol, 98%

Product Code. 15446528
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446528 Supplier Alfa Aesar™ Supplier No. H60061.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
It is a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 120466-66-2
Quantity 1 g
Molecular Formula C8H9ClO
MDL Number MFCD06659510
Synonym r-1-2-chlorophenyl ethanol, 1r-1-2-chlorophenyl ethan-1-ol, 1r-1-2-chlorophenyl-ethan-1-ol, 1r-1-2-chlorophenyl ethanol, r-1-2-chlorophenyl ethan-1-ol, r-2-chloro-alpha-methylbenzyl alcohol, pubchem3980, r-1-2-chlorophenylethanol, r-2'-chloro-1-phenylethanol, r-1-2-chloro-phenyl-ethanol
Solubility Information Slightly soluble in water.
InChI Key DDUBOVLGCYUYFX-ZCFIWIBFSA-N
SMILES CC(C1=CC=CC=C1Cl)O
IUPAC Name (1R)-1-(2-chlorophenyl)ethanol
Molecular Weight (g/mol) 156.609
PubChem CID 6999092
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material (R)-1-(2-Chlorophenyl)ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.