missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Product Code. 15463747
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463747 250 mg 250mg
11382749 1 g 1g
11392749 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15463747 Supplier Thermo Scientific Alfa Aesar Supplier No. H27673.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23357-47-3 | C11H12O2 | 176.215 g/mol

It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23357-47-3
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD07368364
InChI Key VDLWTJCSPSUGOA-SNVBAGLBSA-N
Synonym r-1,2,3,4-tetrahedro-naphthoic acid, r-1,2,3,4-tetrahydronaphthoic acid, r-1,2,3,4-tetrahydro-naphthoic acid, r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, r-1,2,3,4-tetrahydro-1-naphthoic acid, r-1,2,3,4-tetrahedronaphthoic acid, 1r-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, 1-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-, 1r, pubchem5734, r-1,2,3,4-tert-ahydro naphthoic acid
PubChem CID 12598147
IUPAC Name (1R)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 56°C to 58°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Optical Rotation +12° (c=1 in Ethanol)
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (R)-1,2,3,4-Tetrahydro-1-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.