Learn More
(R)-(+)-1,1'-Bi(2-naphthylamine), 97%
Description
(R)-(+)-1,1'-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(R)-(+)-1,1′-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Specifications
| Melting Point | 242°C to 243°C |
| Quantity | 250 mg |
| Beilstein | 5279621 |
| Solubility Information | Slightly soluble in water. |
| Optical Rotation | +150° (c=1 in Pyridine) |
| Formula Weight | 284.36 |
| Percent Purity | 97% |
| Chemical Name or Material | (R)-(+)-1,1'-Bi(2-naphthylamine) |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.