missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,1'-Bi(2-naphthol), 99%

Product Code. 11348095
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348095 1 g 1g
11358095 5 g 5g
11368095 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348095 Supplier Thermo Scientific Alfa Aesar Supplier No. L08305.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18531-94-7 | C20H14O2 | 286.33 g/mol

It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

Solubility
Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50 mg/mL.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18531-94-7
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00004068
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

Specifications

Melting Point 206°C to 211°C
Density 1.301
Odor Characteristic
Quantity 1 g
UN Number UN2811
Beilstein 3616837
Merck Index 14,1226
Solubility Information Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50mg/mL.
Optical Rotation +34.5 (c=1 in THF)
Formula Weight 286.33
Percent Purity 99%
Chemical Name or Material (R)-(+)-1,1'-Bi(2-naphthol)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.