missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ QX 314 bromide

Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Membrane impermeable quaternary derivative of lidocaine, a blocker of voltage-activated Na+ channels. Intracellular QX 314 bromide also inhibits calcium currents in hippocampal CA1 pyramidal neurons.
Specifications
Specifications
| CAS | 24003-58-5 |
| Quantity | 100 mg |
| Molecular Formula | C16H27BrN2O |
| Formula Weight | 343.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 343.309 |
| Chemical Name or Material | QX 314 bromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction