missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quizartinib, MedChemExpress
Description
Quizartinib (AC220) is an orally active, highly selective and potent second-generation type II FLT3 tyrosine kinase inhibitor, with a Kd of 1.6 nM. Quizartinib inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells with IC50s of 4.2 and 1.1 nM, respectively. Quizartinib can be linked to the VHL ligand via an optimized linker to form a PROTAC FLT3 degrader. Quizartinib induces apoptosis.
Specifications
Specifications
| Chemical Name or Material | Quizartinib |
| CAS | 950769-58-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H32N6O4S |
| Quantity | 100 mg |
| Synonym | AC220 |
| Solubility Information | DMSO : ≥ 33 mg/mL (58.86 mM) |
| SMILES | O=C(NC1=NOC(C(C)(C)C)=C1)NC2=CC=C(C=C2)C3=CN4C(SC5=CC(OCCN6CCOCC6)=CC=C45)=N3 |
| Molecular Weight (g/mol) | 560.67 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?