missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quizartinib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Quizartinib (AC220) is an orally active, highly selective and potent second-generation type II FLT3 tyrosine kinase inhibitor, with a Kd of 1.6 nM. Quizartinib inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells with IC50s of 4.2 and 1.1 nM, respectively. Quizartinib can be linked to the VHL ligand via an optimized linker to form a PROTAC FLT3 degrader. Quizartinib induces apoptosis.
Specifications
Specifications
| Chemical Name or Material | Quizartinib |
| CAS | 950769-58-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H32N6O4S |
| Quantity | 5 mg |
| Synonym | AC220 |
| Solubility Information | DMSO : ≥ 33 mg/mL (58.86 mM) |
| SMILES | O=C(NC1=NOC(C(C)(C)C)=C1)NC2=CC=C(C=C2)C3=CN4C(SC5=CC(OCCN6CCOCC6)=CC=C45)=N3 |
| Molecular Weight (g/mol) | 560.67 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction