missing translation for 'onlineSavingsMsg'
Learn More

Quizalofop-p-ethyl, MedChemExpress

Product Code. 16461466
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461466 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461466 Supplier Medchem Express Supplier No. HYB1950200mg
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Quizalofop-P-ethyl is a slightly toxic, selective, postemergence phenoxy herbicide, used to control annual and perennial grass weeds in potatoes, soybeans, sugar beets, peanuts vegetables, cotton and flax.

Quizalofop-P-ethyl is a slightly toxic, selective, postemergence phenoxy herbicide, used to control annual and perennial grass weeds in potatoes, soybeans, sugar beets, peanuts vegetables, cotton and flax.

Specifications

Chemical Name or Material Quizalofop-p-ethyl
CAS 100646-51-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H17ClN2O4
Quantity 200 mg
Synonym (R)-Quizalofop ethyl Quinofop-ethyl
Solubility Information DMSO : ≥ 100 mg/mL (268.24 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@H](OC1=CC=C(OC2=NC3=CC=C(Cl)C=C3N=C2)C=C1)C(OCC)=O
Molecular Weight (g/mol) 372.8
Formula Weight 372.8
Percent Purity 98.84%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.