missing translation for 'onlineSavingsMsg'
Learn More

(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals

Product Code. 15434169
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 15434169

Brand: Thermo Scientific Alfa Aesar J61591.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

Solubility
Soluble in ethanol, water. Insoluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52809-07-1
Molecular Formula C5H7N3O5
Molecular Weight (g/mol) 189.13
MDL Number MFCD00069337
InChI Key ASNFTDCKZKHJSW-REOHCLBHSA-N
Synonym quisqualic acid, quisqualate, l-quisqualic acid, +-quisqualic acid, 3h quisqualate, unii-8oc22c1b99, 2s-2-amino-3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl propanoic acid, 2jbk, 1,2,4-oxadiazolidine-2-propanoic acid, alpha-amino-3,5-dioxo-, s, 3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl-l-alanine
PubChem CID 40539
ChEBI CHEBI:8734
IUPAC Name (2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
SMILES N[C@@H](CN1OC(=O)NC1=O)C(O)=O

Specifications

Melting Point 185°C to 187°C
Color White
Quantity 5 mg
Beilstein 1078734
Merck Index 14,8085
Solubility Information Soluble in ethanol,water. Insoluble in organic solvents.
Formula Weight 189.13
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material (+)-Quisqualic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.