missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline, TRC

Product Code. 30418922
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418922 5 g -
30417564 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418922 Supplier Toronto Research Chemicals Supplier No. TRCQ7500055G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-19-0
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
Synonym 1,4-Benzodiazine, 1,4-Diazanaphthalene, 1,4-Naphthyridine, Benzo[a]pyrazine, Benzoparadiazine, Benzopyrazine, Phenopiazine, Phenpiazine, Quinazine
IUPAC Name quinoxaline
SMILES c1ccc2nccnc2c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H6N2/c1-2-4-8-7(3-1)9-5-6-10-8/h1-6H
Formula Weight 130.0531
Chemical Name or Material Quinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.