missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline-6-carboxylic acid, 95%

Product Code. 15445957
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445957 1 g 1g
15455957 5 g 5g
15465957 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15445957 Supplier Thermo Scientific Alfa Aesar Supplier No. H33157.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6925-00-4 | C9H6N2O2 | 174.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6925-00-4
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD01365834
InChI Key JGQDBVXRYDEWGM-UHFFFAOYSA-N
Synonym 6-quinoxalinecarboxylic acid, chinoxalin-6-carbonsaeure, quinoxaline-6-carboxylicacid, pubchem15419, quinoxaline-6-carboxylic acid
PubChem CID 674813
IUPAC Name quinoxaline-6-carboxylic acid
SMILES C1=CC2=NC=CN=C2C=C1C(=O)O

Specifications

Melting Point 248°C to 251°C
Quantity 1 g
Formula Weight 174.16
Percent Purity 95%
Chemical Name or Material Quinoxaline-6-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.