missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline-2-carboxylic acid, 97%

Product Code. 15426188
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426188 100 mg 100mg
15436188 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426188 Supplier Thermo Scientific Alfa Aesar Supplier No. H59780.MC
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 879-65-2 | C9H6N2O2 | 174.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879-65-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.16
MDL Number MFCD00012334
InChI Key UPUZGXILYFKSGE-UHFFFAOYSA-N
Synonym 2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
PubChem CID 96695
IUPAC Name quinoxaline-2-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC=CC2=N1

Specifications

Melting Point 208°C (decomposition)
Quantity 100 mg
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Quinoxaline-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.