missing translation for 'onlineSavingsMsg'
Learn More

Quinoline, TRC

Product Code. 30424555
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424555 100 g
30410473 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424555 Supplier Toronto Research Chemicals Supplier No. TRCQ700380100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-22-5
Molecular Formula C9 H7 N
Molecular Weight (g/mol) 129.16
Synonym Quinoline, 1-Azanaphthalene, 1-Benzazine, B 500, Benzo[b]pyridine, Benzopyridine, Leucol, Leucoline, Leukol, NSC 3396, Quinolin
IUPAC Name quinoline
SMILES c1ccc2ncccc2c1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H
Formula Weight 129.0578
Chemical Name or Material Quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.