missing translation for 'onlineSavingsMsg'
Learn More

Quinoline N-oxide hydrate, 97% (dry wt.), water ca 20%

Product Code. 11460698
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460698 25 g 25g
11450698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460698 Supplier Thermo Scientific Alfa Aesar Supplier No. B25209.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64201-64-5 | C9H7NO | 145.16 g/mol

Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

Notes
Hygroscopic. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64201-64-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006737
InChI Key GIIWGCBLYNDKBO-UHFFFAOYSA-N
Synonym quinoline n-oxide hydrate, quinoline, 1-oxide, monohydrate, 1-oxidoquinolin-1-ium hydrate, quinoline-n-oxide hydrate, quinoline-1-oxide hydrate, quinolinol, oxamethane, acmc-1be96, quinoline 1-oxide monohydrate, quinoline n-oxide monohydrate
PubChem CID 152363
IUPAC Name 1-oxidoquinolin-1-ium;hydrate
SMILES [O-][N+]1=CC=CC2=CC=CC=C12

Specifications

Melting Point ∼50°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 5989589
Sensitivity Hygroscopic
Formula Weight 145.16 (Anhydrous)
Percent Purity 97%
Assay (dry wt.), water ≈20%
Chemical Name or Material Quinoline N-oxide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.