missing translation for 'onlineSavingsMsg'
Learn More

Quinoline N-oxide hydrate, 97% (dry wt.), water ca 20%

Product Code. 11450698
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11450698

Brand: Thermo Scientific Alfa Aesar B25209.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64201-64-5 | C9H7NO | 145.16 g/mol

Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline N-oxide hydrate is used in quantitative determination of nitrones using trifluoroacetic anhydride-sodium iodide reagent.

Notes
Hygroscopic. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64201-64-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006737
InChI Key GIIWGCBLYNDKBO-UHFFFAOYSA-N
Synonym quinoline n-oxide hydrate, quinoline, 1-oxide, monohydrate, 1-oxidoquinolin-1-ium hydrate, quinoline-n-oxide hydrate, quinoline-1-oxide hydrate, quinolinol, oxamethane, acmc-1be96, quinoline 1-oxide monohydrate, quinoline n-oxide monohydrate
PubChem CID 152363
IUPAC Name 1-oxidoquinolin-1-ium;hydrate
SMILES [O-][N+]1=CC=CC2=CC=CC=C12

Specifications

Melting Point ∼50°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 5989589
Sensitivity Hygroscopic
Formula Weight 145.16 (Anhydrous)
Percent Purity 97%
Assay (dry wt.), water ≈20%
Chemical Name or Material Quinoline N-oxide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.