Learn More
Quinoline, 98%, Thermo Scientific Chemicals
£23.60 - £182.00
Chemical Identifiers
CAS | 91-22-5 |
---|---|
Molecular Formula | C9H7N |
Molecular Weight (g/mol) | 129.16 |
MDL Number | MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 |
InChI Key | SMWDFEZZVXVKRB-UHFFFAOYSA-N |
Synonym | chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol |
PubChem CID | 7047 |
ChEBI | CHEBI:17362 |
IUPAC Name | quinoline |
SMILES | C1=CC=C2N=CC=CC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359298
|
Thermo Scientific Alfa Aesar
043225.18 |
50 g |
£23.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339298
|
Thermo Scientific Alfa Aesar
043225.30 |
250 g |
£64.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349298
|
Thermo Scientific Alfa Aesar
043225.A1 |
1 kg |
£182.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Quinoline acts as a precursor to 8-hydroxyquinoline, hydroxyquinoline sulfate and niacin. It is used as a solvent for resins and terpenes. It is also used in the manufacture of dyes, food colorants, antibiotics, alkaloids, rubber chemicals, flavoring agents, corrosion inhibitors and preservatives. It finds application in transition-metal complex catalysis for uniform polymerization. Further, it is involved in the preparation of pH indicators and other organic compounds. It exhibits antiseptic, antipyretic and antiperiodic properties, thereby it is utilized as antimalarials and for preparing other antimalarial drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-22-5 | |
129.16 | |
SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
7047 | |
quinoline |
C9H7N | |
MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
CHEBI:17362 | |
C1=CC=C2N=CC=CC2=C1 |
Specifications
91-22-5 | |
Colorless to Yellow | |
7.3 | |
101°C (214°F) | |
1.626 | |
UN2656 | |
Hygroscopic; Light sensitive | |
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
quinoline | |
7047 | |
129.16 | |
Liquid |
-15°C | |
1.090 g/mL | |
237°C | |
C9H7N | |
MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
107477 | |
14,8068 | |
Miscible with alcohol,ether and carbon disulfide. Slightly miscible with water. | |
C1=CC=C2N=CC=CC2=C1 | |
129.16 | |
CHEBI:17362 | |
98% | |
Quinoline |
Safety and Handling
GHS H Statement
H311-H350-H341-H302-H315-H319
Toxic in contact with skin.
May cause cancer.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H341-H350
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: QUINOLINE
missing translation for 'einecsNumber' : 202-051-6
missing translation for 'rtecsNumber' : VA9275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only