missing translation for 'onlineSavingsMsg'
Learn More

Quinoline, 96%

Product Code. 10367533 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10367533

Brand: Thermo Scientific Acros 132190025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-22-5 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-22-5
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
InChI Key SMWDFEZZVXVKRB-UHFFFAOYSA-N
Synonym chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
PubChem CID 7047
ChEBI CHEBI:17362
IUPAC Name quinoline
SMILES C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point -15.0°C
Color Brown to Yellow
Density 1.0950g/mL
Boiling Point 237.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6230 to 1.6260
Quantity 2.5 L
Beilstein 20,339
Fieser 01,975; 02,356; 09,403
Merck Index 15,8185
Specific Gravity 1.095
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in hot water,miscible with alcohol,ether,acetone,benzene,and carbon disulfide
Formula Weight 129.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Quinoline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Quinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Quinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.