missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11337545
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11337545

Brand: Thermo Scientific Alfa Aesar L05676.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18704-37-5
Molecular Formula C9H6ClNO2S
Molecular Weight (g/mol) 227.662
MDL Number MFCD00006808
InChI Key JUYUYCIJACTHMK-UHFFFAOYSA-N
Synonym 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp
PubChem CID 29220
IUPAC Name quinoline-8-sulfonyl chloride
SMILES C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2

Specifications

Melting Point 127°C to 130°C
Boiling Point 306°C
Quantity 50 g
UN Number UN3261
Beilstein 156347
Sensitivity Moisture sensitive
Formula Weight 227.67
Percent Purity 97%
Chemical Name or Material Quinoline-8-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.